missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(±)-Benalaxyl-d5 (phenyl-d5), CDN
missing translation for 'orderingAttributeHoverText'
Quantity:
0.01 g
0.05 g
missing translation for 'unitSize'
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | Benalaxyl-d5 |
| Molecular Formula | C20 D5 H18 N O3 |
| Quantity | 0.01 g |
| Synonym | (±)-Benalaxyl-d5, Benalaxyl-D5 (phenyl-D5), Methyl 2-(N-(2,6-dimethylphenyl)-2-(phenyl-d5)acetamido)propanoate, N-(2,6-Dimethylphenyl)-N-[2-(phenyl-d5)acetyl]-DL-alanine methyl ester |
| InChI Formula | InChI=1 S/C20H23NO3/c1-14-9-8-10-15(2)19(14)21(16(3)20(23)24-4)18(22)13-17-11-6-5-7-12-17/h5-12,16 H,13H2,1-4H3/i5D,6 D,7 D,11 D,12 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(CC(=O)N(C(C)C(=O)OC)c2c(C)cccc2C)c([2 H])c1[2 H] |
| IUPAC Name | methyl 2-(2,6-dimethyl-N-[2-(2,3,4,5,6-pentadeuteriophenyl)acetyl]anilino)propanoate |
| Molecular Weight (g/mol) | 330.4323 |
| Formula Weight | 330.1992 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction