Learn More
Atrazine-d5 (ethyl-d5), CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | Atrazine-d5 |
| CAS | 163165-75-1 |
| Molecular Formula | C8 2H5 H9 Cl N5 |
| Quantity | 0.01 g |
| Synonym | 1,3,5-Triazine-2,4-diamine, 6-chloro-N-(ethyl-d5)-N'-(1-methylethyl)- (9 CI), Atrazine D5 (ethyl-D5), Atrazine D5, Atrazine-d5, Atrazine D5 (ethylamino D5) |
| InChI Formula | InChI=1 S/C8H14ClN5/c1-4-10-7-12-6(9)13-8(14-7)11-5(2)3/h5H,4H2,1-3H3,(H2,10,11,12,13,14)/i1D3,4D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])Nc1nc(Cl)nc(NC(C)C)n1 |
| IUPAC Name | 6-chloro-4-N-(1,1,2,2,2-pentadeuterioethyl)-2-N-propan-2-yl-1,3,5-triazine-2,4-diamine |
| Molecular Weight (g/mol) | 220.71 |
| Formula Weight | 220.1252 g/mol |
| Show More |