missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(±)-Atomoxetine-d5 HCl (phenyl-d5), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | rac Atomoxetine-d5 Hydrochloride |
| CAS | 1398065-95-6 |
| Molecular Formula | C17 D5 H16 N O . Cl H |
| Quantity | 0.005 g |
| Synonym | N-Methyl-3-(pentadeuteriophenyl)-3-(2-methylphenoxy)propan-1-amine hydrochloride (1:1), 3-(2-Methylphenoxy)-3-(phenyl-d5)-N-methylpropan-1-amine hydrochloride, N-Methyl-3-(2-methylphenoxy)-3-(phenyl-d5)propylamine monohydrochloride, rac Atomoxetine-d5 hydrochloride, (±)-Atomoxetine-d5 hydrochloride, (RS)-Atomoxetine-D5 (phenyl-D5) hydrochloride |
| InChI Formula | InChI=1 S/C17H21NO.ClH/c1-14-8-6-7-11-16(14)19-17(12-13-18-2)15-9-4-3-5-10-15;/h3-11,17-18 H,12-13H2,1-2H3;1 H/i3D,4 D,5 D,9 D,10 D; |
| SMILES | Cl.[2 H]c1c([2 H])c([2 H])c(C(CCNC)Oc2ccccc2C)c([2 H])c1[2 H] |
| IUPAC Name | N-methyl-3-(2-methylphenoxy)-3-(2,3,4,5,6-pentadeuteriophenyl)propan-1-amine;hydrochloride |
| Molecular Weight (g/mol) | 296.846 |
| Formula Weight | 296.17 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction