missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5beta-Pregnan-3alpha,21-diol-20-one-17alpha,21,21-d3, CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | 5 beta-Pregnan-3 alpha,21-diol-20-one-17 alpha,21,21-d3 |
| CAS | 72205-58-4 |
| Molecular Formula | C21 D3 H31 O3 |
| Quantity | 0.01 g |
| Synonym | Pregnan-20-one-17,21,21-d3, 3,21-dihydroxy-, (3α,5β)- (9 CI), 5β-Pregnan-3α,21-diol-20-one-17α,21,21-d3, (3α,5β)-3,21-Dihydroxypregnan-20-one-17α,21,21-d3, 3α,21-Dihydroxy-5β-pregnan-20-one-17,21,21-D3, Tetrahydro-11-deoxycorticosterone-17,21,21-d3 |
| InChI Formula | InChI=1 S/C21H34O3/c1-20-9-7-14(23)11-13(20)3-4-15-16-5-6-18(19(24)12-22)21(16,2)10-8-17(15)20/h13-18,22-23 H,3-12H2,1-2H3/t13-,14-,15+,16+,17+,18-,20+,21+/m1/s1/i12D2,18 D |
| SMILES | [2 H]C([2 H])(O)C(=O)[C@@]1([2 H])CC[C@H]2 [C@@H]3 CC[C@@H]4 C[C@H](O)CC[C@]4(C)[C@H]3 CC[C@]12 C |
| IUPAC Name | 2,2-dideuterio-1-[(3 R,5 R,8 R,9 S,10 S,13 S,14 S,17 S)-17-deuterio-3-hydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,14,15,16-tetradecahydrocyclopenta[a]phenanthren-17-yl]-2-hydroxyethanone |
| Molecular Weight (g/mol) | 337.5114 |
| Formula Weight | 337.2696 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction