missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5beta-Androstan-3alpha-ol-17-one-16,16-d2, CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | 5 beta-Androsterone-16,16-D2 |
| CAS | 2687960-82-1 |
| Molecular Formula | C19 D2 H28 O2 |
| Quantity | 0.01 g |
| Synonym | Androstan-17-one-16,16-d2, 3-hydroxy-, (3α,5β)- (ACI), (3α,5β)-3-Hydroxyandrostan-17-one-16,16-d2 (ACI), 5β-Androstan-3α-ol-17-one-16,16-d2, Etiocholanolone-16,16-d2, 5β-Androsterone-16,16-d2, Etiocholan-3α-ol-17-one-16,16-d2, 3α-Etiocholanolone-16,16-d2, 5β-Androsterone-16,16-D2 |
| InChI Formula | InChI=1 S/C19H30O2/c1-18-9-7-13(20)11-12(18)3-4-14-15-5-6-17(21)19(15,2)10-8-16(14)18/h12-16,20 H,3-11H2,1-2H3/t12-,13-,14+,15+,16+,18+,19+/m1/s1/i6D2 |
| SMILES | [2 H]C1([2 H])C[C@H]2 [C@@H]3 CC[C@@H]4 C[C@H](O)CC[C@]4(C)[C@H]3 CC[C@]2(C)C1=O |
| IUPAC Name | (3 R,5 R,8 R,9 S,10 S,13 S,14 S)-16,16-dideuterio-3-hydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15-dodecahydro-1 H-cyclopenta[a]phenanthren-17-one |
| Molecular Weight (g/mol) | 292.4526 |
| Formula Weight | 292.2371 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction