missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5alpha-Pregnan-3alpha-ol-20-one-17alpha,21,21,21-d4, CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | 3 alpha-Hydroxy-5 alpha-pregnan-20-one-17,21,21,21-D4 |
| CAS | 203805-85-0 |
| Molecular Formula | C21 H30 D4 O2 |
| Quantity | 0.005 g |
| Synonym | Pregnan-20-one-17,21,21,21-d4, 3-hydroxy-, (3α,5α)- (9 CI, ACI), (3α,5α)-3-Hydroxypregnan-20-one-17,21,21,21-d4 (ACI), (3α)-Allopregnanolone-d4, Brexanolone-17,21,21,21-d4, 3α-Hydroxy-5α-pregnan-20-one-17,21,21,21-D4, 5 alpha-Pregnan-3 alpha-ol-20-one-17 alpha,21,21,21-d4, 5 alpha-Pregnan-3 alpha-ol-20-one-17 alpha,21,21,21-d4, 5 alpha-Pregnan-3 alpha-ol-20-one-17 alpha,21,21,21-d4, 5 alpha-Pregnan-3 alpha-ol-20-one-17 alpha,21,21,21-d4, 5 alpha-Pregnan-3 alpha-ol-20-one-17 alpha,21,21,21-d4, 5 alpha-Pregnan-3 alpha-ol-20-one-17 alpha,21,21,21-d4 |
| InChI Formula | InChI=1 S/C21H34O2/c1-13(22)17-6-7-18-16-5-4-14-12-15(23)8-10-20(14,2)19(16)9-11-21(17,18)3/h14-19,23 H,4-12H2,1-3H3/t14-,15+,16-,17+,18-,19-,20-,21+/m0/s1/i1D3,17 D |
| SMILES | C[C@@]12 [C@@]3([C@]([C@]4([C@](C)(CC3)[C@@](C(C([2 H])([2 H])[2 H])=O)(CC4)[2 H])[H])(CC[C@]1(C[C@H](O)CC2)[H])[H])[H] |
| IUPAC Name | 2,2,2-trideuterio-1-[(3 R,5 S,8 R,9 S,10 S,13 S,14 S,17 S)-17-deuterio-3-hydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,14,15,16-tetradecahydrocyclopenta[a]phenanthren-17-yl]ethanone |
| Molecular Weight (g/mol) | 322.5181 |
| Formula Weight | 322.281 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction