Learn More
5alpha-Cholanic Acid-3beta-ol-23,23-d2, CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Isoallolithocholic Acid-23,23-D2 |
| CAS | 2410277-69-7 |
| Molecular Formula | C24 D2 H38 O3 |
| Quantity | 0.01 g |
| Synonym | Cholan-24-oic-23,23-d2 acid, 3-hydroxy-, (3β,5α)- (ACI), (3β,5α)-3-Hydroxycholan-24-oic-23,23-d2 acid (ACI), 5α-Cholanic Acid-3β-ol-23,23-d2, 3β-Hydroxy-5α-cholanic acid-23,23-d2, 3β-Hydroxy-5α-cholanoic-23,23-d2 acid, Isoallolithocholic acid-23,23-d2, Isoallolithocholic Acid-23,23-D2 |
| InChI Formula | InChI=1 S/C24H40O3/c1-15(4-9-22(26)27)19-7-8-20-18-6-5-16-14-17(25)10-12-23(16,2)21(18)11-13-24(19,20)3/h15-21,25 H,4-14H2,1-3H3,(H,26,27)/t15-,16+,17+,18+,19-,20+,21+,23+,24-/m1/s1/i9D2 |
| SMILES | [2 H]C([2 H])(C[C@@H](C)[C@H]1 CC[C@H]2 [C@@H]3 CC[C@H]4 C[C@@H](O)CC[C@]4(C)[C@H]3 CC[C@]12 C)C(=O)O |
| IUPAC Name | (4 R)-2,2-dideuterio-4-[(3 S,5 S,8 R,9 S,10 S,13 R,14 S,17 R)-3-hydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1 H-cyclopenta[a]phenanthren-17-yl]pentanoic acid |
| Molecular Weight (g/mol) | 378.5849 |
| Formula Weight | 378.3103 g/mol |
| Show More |