missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5alpha-Androstan-3beta,17beta-diol-16,16,17-d3, CDN
Click to view available options
Quantity:
0.005 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | d3-5 A-Androstane-3 b,17 b-diol |
| CAS | 79037-32-4 |
| Molecular Formula | C19 2H3 H29 O2 |
| Quantity | 0.005 g |
| SMILES | [2 H]C1([2 H])C[C@H]2 [C@@H]3 CC[C@H]4 C[C@@H](O)CC[C@]4(C)[C@H]3 CC[C@]2(C)[C@@]1([2 H])O |
| IUPAC Name | (3 S,5 S,8 R,9 S,10 S,13 S,14 S,17 S)-16,16,17-trideuterio-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15-dodecahydro-1 H-cyclopenta[a]phenanthrene-3,17-diol |
| Molecular Weight (g/mol) | 295.4 |
| Formula Weight | 295.4747 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction