Learn More
5-Methyl-d3-cytosine-6-d1, CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | 5-Methylcytosine methyl,6-d4 |
| CAS | 1219795-15-9 |
| Molecular Formula | C5 2H4 H3 N3 O |
| Quantity | 0.01 g |
| Synonym | 6-Amino-5-(methyl-d3)-2(1 H)-pyrimidinone-4-d, 4-Amino-2-hydroxy-5-(methyl-d3)pyrimidine-6-d, 4-Amino-5-(methyl-d3)-2(1 H)-pyrimidinone-6-d, 5-Methylcytosine-d4, 5-Methylcytosine-D4 (methyl-D3,6-D) |
| InChI Formula | InChI=1 S/C5H7N3O/c1-3-2-7-5(9)8-4(3)6/h2H,1H3,(H3,6,7,8,9)/i1D3,2 D |
| SMILES | [2 H]C1=C(C(=NC(=O)N1)N)C([2 H])([2 H])[2 H] |
| IUPAC Name | 4-amino-6-deuterio-5-(trideuteriomethyl)-1 H-pyrimidin-2-one |
| Molecular Weight (g/mol) | 129.15 |
| Formula Weight | 129.084 g/mol |
| Show More |