missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Pregnen-11beta,17alpha-diol-3,20-dione-2,2,4,6,6,21,21,21-d8, CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | 11 beta,17-Dihydroxypregn-4-ene-3,20-dione-2,2,4,6,6,21,21,21-D8 |
| CAS | 2479914-04-8 |
| Molecular Formula | C21 D8 H22 O4 |
| Quantity | 0.01 g |
| Synonym | Pregn-4-ene-3,20-dione-2,2,4,6,6,21,21,21-d8, 11,17-dihydroxy-, (11β)- (ACI), (11β)-11,17-Dihydroxypregn-4-ene-3,20-dione-2,2,4,6,6,21,21,21-d8 (ACI), 21-Deoxycortisol-D8 (2,2,4,6,6,21,21,21-D8), 21-Deoxycortisol-2,2,4,6,6,21,21,21-D8, 21-Deoxycortisol-d8, 11β,17-Dihydroxypregn-4-ene-3,20-dione-2,2,4,6,6,21,21,21-d8, 11β,17α-Dihydroxyprogesterone-2,2,4,6,6,21,21,21-d8, 21-Dehydrohydrocortisone-2,2,4,6,6,21,21,21-d8, 11β,17α-Dihydroxyprogesterone-d8, 21-Dehydrohydrocortisone-d8 |
| InChI Formula | InChI=1 S/C21H30O4/c1-12(22)21(25)9-7-16-15-5-4-13-10-14(23)6-8-19(13,2)18(15)17(24)11-20(16,21)3/h10,15-18,24-25 H,4-9,11H2,1-3H3/t15-,16-,17-,18+,19-,20-,21-/m0/s1/i1D3,4D2,6D2,10 D |
| SMILES | [2 H]C1=C2C([2 H])([2 H])C[C@H]3 [C@@H]4 CC[C@](O)(C(=O)C([2 H])([2 H])[2 H])[C@@]4(C)C[C@H](O)[C@@H]3 [C@@]2(C)CC([2 H])([2 H])C1=O |
| IUPAC Name | (8 S,9 S,10 R,11 S,13 S,14 S,17 R)-2,2,4,6,6-pentadeuterio-11,17-dihydroxy-10,13-dimethyl-17-(2,2,2-trideuterioacetyl)-7,8,9,11,12,14,15,16-octahydro-1 H-cyclopenta[a]phenanthren-3-one |
| Molecular Weight (g/mol) | 354.5098 |
| Formula Weight | 354.2646 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction