Learn More
3-Nitrofluoranthene-d9, CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | 3-Nitrofluoranthene (D9) |
| CAS | 350820-11-0 |
| Molecular Formula | C16 D9 N O2 |
| Quantity | 0.01 g |
| Synonym | Fluoranthene-1,2,3,5,6,7,8,9,10-d9, 4-nitro- (9 CI), 3-Nitrofluoranthene-d9, 3-Nitrofluoranthene-D9, 4-Nitrofluoranthene-d9, 4-Nitrofluoranthene-1,2,3,5,6,7,8,9,10-d9 |
| InChI Formula | InChI=1 S/C16H9NO2/c18-17(19)15-9-8-13-11-5-2-1-4-10(11)12-6-3-7-14(15)16(12)13/h1-9 H/i1D,2 D,3 D,4 D,5 D,6 D,7 D,8 D,9 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c2c(c1[2 H])c3c([2 H])c([2 H])c([2 H])c4c(c([2 H])c([2 H])c2c34)[N+](=O)[O-] |
| IUPAC Name | 1,2,3,5,6,7,8,9,10-nonadeuterio-4-nitrofluoranthene |
| Molecular Weight (g/mol) | 256.3036 |
| Formula Weight | 256.1198 g/mol |
| Show More |