Learn More
3-Mercaptopropionic-2,2,3,3-d4 Acid, CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | 3-Thiopropanoic Acid-D4 |
| CAS | 1141488-67-6 |
| Molecular Formula | C3 D4 H2 O2 S |
| Quantity | 0.01 g |
| Synonym | Propanoic-2,2,3,3-d4 acid, 3-mercapto- (ACI), 3-Mercaptopropanoic-2,2,3,3-d4 acid (ACI), 3-Mercaptopropanoic acid-2,2,3,3-d4, 3-Mercaptopropanoic acid-d4, 3-Thiopropanoic acid-2,2,3,3-d4, 3-Thiopropanoic acid-D4, 3-Mercaptopropionic-2,2,3,3-d4 acid |
| InChI Formula | InChI=1 S/C3H6O2S/c4-3(5)1-2-6/h6H,1-2H2,(H,4,5)/i1D2,2D2 |
| SMILES | [2 H]C([2 H])C([2 H])([2 H])C(=O)O |
| IUPAC Name | 2,2,3,3-tetradeuterio-3-sulfanylpropanoic acid |
| Molecular Weight (g/mol) | 110.1682 |
| Formula Weight | 110.034 g/mol |
| Show More |