Learn More
3,5-Dimethoxy-d6-4-hydroxybenzoic Acid, CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Syringic-d6 Acid |
| CAS | 84759-06-8 |
| Molecular Formula | C9 D6 H4 O5 |
| Quantity | 0.005 g |
| Synonym | Benzoic acid, 4-hydroxy-3,5-di(methoxy-d3)- (9 CI), 3,5-Dimethoxy-d6-4-hydroxybenzoic Acid, Syringic-d6 Acid, Syringic Acid-D6 (dimethoxy-D6), 2,6-Di(methoxy-d3)-4-carboxyphenol, 3,5-Di(methoxy-d3)-4-hydroxybenzoic acid, Gallic acid 3,5-di(methyl-d3) ether |
| InChI Formula | InChI=1 S/C9H10O5/c1-13-6-3-5(9(11)12)4-7(14-2)8(6)10/h3-4,10 H,1-2H3,(H,11,12)/i1D3,2D3 |
| SMILES | [2 H]C([2 H])([2 H])Oc1cc(cc(OC([2 H])([2 H])[2 H])c1O)C(=O)O |
| IUPAC Name | 4-hydroxy-3,5-bis(trideuteriomethoxy)benzoic acid |
| Molecular Weight (g/mol) | 204.2097 |
| Formula Weight | 204.0905 g/mol |
| Show More |