Learn More
3,3'-Dichlorobenzidine-d6 (rings-d6), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | 3,3'-DichlorobenzidineD6(ringD6) |
| CAS | 93951-91-8 |
| Molecular Formula | C12 2H6 H4 Cl2 N2 |
| Quantity | 0.005 g |
| Synonym | [1,1'-Biphenyl-2,2',3,3',6,6'-d6]-4,4'-diamine, 5,5'-dichloro- (9 CI), 3,3'-Dichlorobenzidine D6 (ring D6), 3,3'-Dichlorobenzidine-d6 |
| InChI Formula | InChI=1 S/C12H10Cl2N2/c13-9-5-7(1-3-11(9)15)8-2-4-12(16)10(14)6-8/h1-6 H,15-16H2/i1D,2 D,3 D,4 D,5 D,6 D |
| SMILES | [2 H]c1c([2 H])c(c([2 H])c(Cl)c1N)c2c([2 H])c([2 H])c(N)c(Cl)c2[2 H] |
| IUPAC Name | 4-(4-amino-3-chloro-2,5,6-trideuteriophenyl)-2-chloro-3,5,6-trideuterioaniline |
| Molecular Weight (g/mol) | 259.16 |
| Formula Weight | 258.0598 g/mol |
| Show More |