missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,3',4,4'-Tetrachlorobiphenyl-d6, CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | 3,3',4,4'-Tetrachlorobiphenyl-d6 |
| CAS | 93952-23-9 |
| Molecular Formula | C12 D6 Cl4 |
| Quantity | 0.01 g |
| Synonym | 1,1'-Biphenyl-2,2',3,3',6,6'-d6, 4,4',5,5'-tetrachloro- (9 CI), 3,3',4,4'-Tetrachlorobiphenyl-2,2',5,5',6,6'-d6, PCB 77-d6, PCB 77 d6; |
| InChI Formula | InChI=1 S/C12H6Cl4/c13-9-3-1-7(5-11(9)15)8-2-4-10(14)12(16)6-8/h1-6 H/i1D,2 D,3 D,4 D,5 D,6 D |
| SMILES | [2 H]c1c([2 H])c(c([2 H])c(Cl)c1Cl)c2c([2 H])c([2 H])c(Cl)c(Cl)c2[2 H] |
| IUPAC Name | 1,2-dichloro-3,4,6-trideuterio-5-(3,4-dichloro-2,5,6-trideuterio-phenyl)benzene |
| Molecular Weight (g/mol) | 298.03 |
| Formula Weight | 295.96 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction