missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(2S,2'S)-Ethambutol-d10 (1,1,1',1',2,2'-d6; ethylene-d4), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Ethambutol-D10 (bis[1-butanol-1,1,2-D3],ethylene-D4) |
| CAS | 1129526-24-4 |
| Molecular Formula | C10 D10 H14 N2 O2 |
| Quantity | 0.01 g |
| Synonym | 1-Butan-1,1,2-d3-ol, 2,2'-(1,2-ethanediyl-1,1,2,2-d4-diimino)bis-, (2 S,2'S)- (ACI), (2 S,2'S)-2,2'-(1,2-Ethanediyl-1,1,2,2-d4-diimino)bis[1-butan-1,1,2-d3-ol] (ACI), (2 S,2'S)-Ethambutol-d10 (1,1,1',1',2,2'-d6), ethylene-d4), Ethambutol-d10, Ethambutol-D10 (bis[1-butanol-1,1,2-D3],ethylene-D4) |
| InChI Formula | InChI=1 S/C10H24N2O2/c1-3-9(7-13)11-5-6-12-10(4-2)8-14/h9-14 H,3-8H2,1-2H3/t9-,10-/m0/s1/i5D2,6D2,7D2,8D2,9 D,10 D |
| SMILES | [2 H]C([2 H])(O)[C@]([2 H])(CC)NC([2 H])([2 H])C([2 H])([2 H])N[C@@]([2 H])(CC)C([2 H])([2 H])O |
| IUPAC Name | (2 S)-1,1,2-trideuterio-2-[[1,1,2,2-tetradeuterio-2-[[(2 S)-1,1,2-trideuterio-1-hydroxybutan-2-yl]amino]ethyl]amino]butan-1-ol |
| Molecular Weight (g/mol) | 214.3714 |
| Formula Weight | 214.2465 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction