Learn More
2-Methoxy-17beta-estradiol-1,4,16,16,17-d5, CDN
Click to view available options
Quantity:
0.005 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | 2-Methoxyestradiol-(1,4,16,16,17-D5) |
| CAS | 358731-34-7 |
| Molecular Formula | C19 D4 H22 O3 |
| Quantity | 0.005 g |
| Synonym | Estra-1,3,5(10)-triene-1,4,16,16,17-d5-3,17-diol, 2-methoxy-, (17β)-, (17β)-2-Methoxyestra-1,3,5(10)-triene-1,4,16,16,17-d5-3,17-diol, 2-Methoxyestradiol-(1,4,16,16,17-D5) |
| InChI Formula | InChI=1 S/C19H26O3/c1-19-8-7-12-13(15(19)5-6-18(19)21)4-3-11-9-16(20)17(22-2)10-14(11)12/h9-10,12-13,15,18,20-21 H,3-8H2,1-2H3/t12-,13+,15-,18-,19-/m0/s1/i6D2,9 D,10 D |
| SMILES | [2 H]c1c(O)c(OC)c([2 H])c2[C@H]3 CC[C@@]4(C)[C@@H](CC([2 H])([2 H])[C@@H]4 O)[C@@H]3 CCc12 |
| IUPAC Name | (8 R,9 S,13 S,14 S,17 S)-1,4,16,16-tetradeuterio-2-methoxy-13-methyl-7,8,9,11,12,14,15,17-octahydro-6 H-cyclopenta[a]phenanthrene-3,17-diol |
| Molecular Weight (g/mol) | 306.4326 |
| Formula Weight | 306.2133 g/mol |
| Show More |