missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Di-(tert-butyl-d9)-4-methoxyphenol-3,5-d2, CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | Dibutylated Hydroxyanisole-d20 |
| CAS | 1219799-34-4 |
| Molecular Formula | C15 2H20 H4 O2 |
| Quantity | 0.01 g |
| InChI Formula | InChI=1 S/C15H24O2/c1-14(2,3)11-8-10(17-7)9-12(13(11)16)15(4,5)6/h8-9,16 H,1-7H3/i1D3,2D3,3D3,4D3,5D3,6D3,8 D,9 D |
| SMILES | [2 H]c1c(OC)c([2 H])c(c(O)c1C(C([2 H])([2 H])[2 H])(C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H])C(C([2 H])([2 H])[2 H])(C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | 3,5-dideuterio-2,6-bis[1,1,1,3,3,3-hexadeuterio-2-(trideuteriomethyl)propan-2-yl]-4-methoxyphenol |
| Molecular Weight (g/mol) | 256.47 |
| Formula Weight | 256.3032 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction