Learn More
2,3,4,4',5-Pentachlorobiphenyl-2',3',5',6'-d4, CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | 2,3,4,4',5-Pentachlorobiphenyl-2',3',5',6'-D4 |
| CAS | 1219799-24-2 |
| Molecular Formula | C12 2H4 H Cl5 |
| Quantity | 0.01 g |
| Synonym | 2,3,4,4',5-Pentachlorobiphenyl-2',3',5',6'-D4, 2',3',5',6'-D4-2,3,4,4',5-PeCB, PCB 114-d4, 2,3,4,4',5-Pentachloro-1,1'-biphenyl l-2',3',5',6'-d4, 2,3,4,4',5-Pentachlorobiphenyl D4, 2,3,4,4',5-PentaCB--d4, 2,3,4,4',5-PentaCB--d4, 2,3,4,4',5-PentaCB--d4, 2,3,4,4',5-PentaCB--d4, 2,3,4,4',5-PentaCB--d4 |
| InChI Formula | InChI=1 S/C12H5Cl5/c13-7-3-1-6(2-4-7)8-5-9(14)11(16)12(17)10(8)15/h1-5 H/i1D,2 D,3 D,4 D |
| SMILES | [2 H]c1c([2 H])c(c([2 H])c([2 H])c1Cl)c2cc(Cl)c(Cl)c(Cl)c2Cl |
| IUPAC Name | 1-chloro-2,3,5,6-tetradeuterio-4-(2,3,4,5-tetrachlorophenyl)benzene |
| Molecular Weight (g/mol) | 330.46 |
| Formula Weight | 327.9085 g/mol |
| Show More |