missing translation for 'onlineSavingsMsg'
Learn More
Learn More
17beta-Estradiol-2,4-d2, CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | 17 beta-Estradiol D2 (2,4 D2) |
| CAS | 53866-33-4 |
| Molecular Formula | C18 D2 H22 O2 |
| Quantity | 0.01 g |
| Synonym | 17 beta-Estradiol D2 (2,4-D2), Estra-1,3,5(10)-triene-2,4-d2-3,17-diol, (17β)-, (17β)-Estra-1,3,5(10)-triene-2,4-d2-3,17-diol, 2,4-Dideuterioestradiol: 17 beta-Estradiol D2 (2,4 D2) |
| InChI Formula | InChI=1 S/C18H24O2/c1-18-9-8-14-13-5-3-12(19)10-11(13)2-4-15(14)16(18)6-7-17(18)20/h3,5,10,14-17,19-20 H,2,4,6-9H2,1H3/t14-,15-,16+,17+,18+/m1/s1/i3D,10 D |
| SMILES | [2 H]c1cc2[C@H]3 CC[C@]4(C)[C@@H](O)CC[C@H]4 [C@@H]3 CCc2c([2 H])c1O |
| IUPAC Name | (8 R,9 S,13 S,14 S,17 S)-2,4-dideuterio-13-methyl-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthrene-3,17-diol |
| Molecular Weight (g/mol) | 274.39 |
| Formula Weight | 274.1902 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction