Learn More
16beta-Hydroxy-17beta-estradiol-2,4-d2, CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | 16-epi-Estriol-2,4-D2 |
| CAS | 366495-94-5 |
| Molecular Formula | C18 D2 H22 O3 |
| Quantity | 0.01 g |
| Synonym | Estra-1,3,5(10)-triene-2,4-d2-3,16,17-triol, (16β,17β)- (9 CI), (16β,17β)-Estra-1,3,5(10)-triene-3,16,17-triol-2,4-d2, 16-epi-Estriol-2,4-d2, 16-epi-Estriol-2,4-D2, 16β,17β-Estriol-2,4-d2, 16β-Hydroxyestradiol-2,4-d2, 16β-Hydroxy-17β-estradiol-2,4-d2 |
| InChI Formula | InChI=1 S/C18H24O3/c1-18-7-6-13-12-5-3-11(19)8-10(12)2-4-14(13)15(18)9-16(20)17(18)21/h3,5,8,13-17,19-21 H,2,4,6-7,9H2,1H3/t13-,14-,15+,16+,17+,18+/m1/s1/i3D,8 D |
| SMILES | [2 H]c1cc2[C@H]3 CC[C@]4(C)[C@@H](O)[C@@H](O)C[C@H]4 [C@@H]3 CCc2c([2 H])c1O |
| IUPAC Name | (8 R,9 S,13 S,14 S,16 S,17 R)-2,4-dideuterio-13-methyl-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthrene-3,16,17-triol |
| Molecular Weight (g/mol) | 290.3937 |
| Formula Weight | 290.1851 g/mol |
| Show More |