Learn More
1-Nitropyrene-d9, CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | 1-Nitropyrene-d9 |
| CAS | 93487-20-8 |
| Molecular Formula | C16 2H9 N O2 |
| Quantity | 0.01 g |
| Synonym | Pyrene-1,2,3,4,5,6,7,9,10-d9, 8-nitro-, 8-Nitropyrene-1,2,3,4,5,6,7,9,10-d9, 1-Nitropyrene-d9, 1-Nitropyrene D9 |
| InChI Formula | InChI=1 S/C16H9NO2/c18-17(19)14-9-7-12-5-4-10-2-1-3-11-6-8-13(14)16(12)15(10)11/h1-9 H/i1D,2 D,3 D,4 D,5 D,6 D,7 D,8 D,9 D |
| SMILES | [2 H]c1c([2 H])c2c([2 H])c([2 H])c3c([2 H])c([2 H])c(c4c([2 H])c([2 H])c(c1[2 H])c2c34)[N+](=O)[O-] |
| IUPAC Name | 1,2,3,4,5,6,7,9,10-nonadeuterio-8-nitropyrene |
| Molecular Weight (g/mol) | 256.3 |
| Formula Weight | 256.1198 g/mol |
| Show More |