Learn More
1-Aminoadamantane-d15, CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | Amantadine-d15 |
| CAS | 33830-10-3 |
| Molecular Formula | C10 2H15 H2 N |
| Quantity | 0.01 g |
| InChI Formula | InChI=1 S/C10H17N/c11-10-4-7-1-8(5-10)3-9(2-7)6-10/h7-9 H,1-6,11H2/t7-,8+,9-,10-/i1D2,2D2,3D2,4D2,5D2,6D2,7 D,8 D,9 D |
| SMILES | [2 H]C1([2 H])[C@@]2([2 H])C([2 H])([2 H])[C@@]3([2 H])C([2 H])([2 H])[C@]1([2 H])C([2 H])([2 H])[C@](N)(C3([2 H])[2 H])C2([2 H])[2 H] |
| IUPAC Name | 2,2,3,4,4,5,6,6,7,8,8,9,9,10,10-pentadecadeuterioadamantan-1-amine |
| Molecular Weight (g/mol) | 166.34 |
| Formula Weight | 166.2303 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |