missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Pregnadien-6alpha-methyl-11beta,17alpha,21-triol-3,20-dione-21,21-d2, CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Methylprednisolone-21,21-D2 |
| Molecular Formula | C22 D2 H28 O5 |
| Quantity | 0.01 g |
| Synonym | 1,4-Pregnadien-6α-methyl-11β,17α,21-triol-3,20-dione-21,21-d2, (6α,11β)-11,17,21-Trihydroxy-6-methylpregna-1,4-diene-3,20-dione-21,21-d2, Methylprednisolone-d2, Methylprednisolone-21,21-D2, 11β,17α,21-Trihydroxy-6α-methylpregnane-1,4-diene-3,20-dione-21,21-d2 |
| InChI Formula | InChI=1 S/C22H30O5/c1-12-8-14-15-5-7-22(27,18(26)11-23)21(15,3)10-17(25)19(14)20(2)6-4-13(24)9-16(12)20/h4,6,9,12,14-15,17,19,23,25,27 H,5,7-8,10-11H2,1-3H3/t12-,14-,15-,17-,19+,20-,21-,22-/m0/s1/i11D2 |
| SMILES | [2 H]C([2 H])(O)C(=O)[C@@]1(O)CC[C@H]2 [C@@H]3 C[C@H](C)C4=CC(=O)C=C[C@]4(C)[C@H]3 [C@@H](O)C[C@]12 C |
| IUPAC Name | (6 S,8 S,9 S,10 R,11 S,13 S,14 S,17 R)-17-(2,2-dideuterio-2-hydroxyacetyl)-11,17-dihydroxy-6,10,13-trimethyl-7,8,9,11,12,14,15,16-octahydro-6 H-cyclopenta[a]phenanthren-3-one |
| Molecular Weight (g/mol) | 376.4829 |
| Formula Weight | 376.2219 g/mol |
| Percent Purity | 95 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction