missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(2-Phenyl-d5-ethyl)biguanide HCl, CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Phenformin-d5 Hydrochloride |
| Molecular Formula | C10 D5 H10 N5 . Cl H |
| Quantity | 0.01 g |
| Synonym | N-[2-(Phenyl-2,3,4,5,6-d5)ethyl]imidodicarbonimidic diamide hydrochloride, 1-[2-(Phenyl-d5)ethyl]biguanide hydrochloride, Phenformin-d5 hydrochloride, Phenformin-D5 (phenyl-D5) hydrochloride, 1-(2-Phenyl-d5-ethyl)biguanide HCl |
| InChI Formula | InChI=1 S/C10H15N5.ClH/c11-9(12)15-10(13)14-7-6-8-4-2-1-3-5-8;/h1-5 H,6-7H2,(H6,11,12,13,14,15);1 H/i1D,2 D,3 D,4 D,5 D; |
| SMILES | Cl.[2 H]c1c([2 H])c([2 H])c(CCNC(=N)NC(=N)N)c([2 H])c1[2 H] |
| IUPAC Name | 1-carbamimidoyl-3-[2-(2,3,4,5,6-pentadeuteriophenyl)ethyl]guanidine;hydrochloride |
| Molecular Weight (g/mol) | 246.751 |
| Formula Weight | 246.141 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction