Learn More
1,1'-Sulfonylbis[2-(methyl-d3-thio)ethane], CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | 1,1'-Sulfonylbis[2-(methylthio)ethane]-D6 (S,S-dimethyl-D6) |
| CAS | 2733162-45-1 |
| Molecular Formula | C6 D6 H8 O2 S3 |
| Quantity | 0.01 g |
| Synonym | 1,1'-Sulfonylbis[2-(methylthio-d3)ethane], 1,1'-Sulfonylbis[2-(methyl-d3-thio)ethane], 1,1'-Sulfonylbis[2-(methylthio)ethane]-D6 (S,S-dimethyl-D6) |
| InChI Formula | InChI=1 S/C6H14O2S3/c1-9-3-5-11(7,8)6-4-10-2/h3-6H2,1-2H3/i1D3,2D3 |
| SMILES | [2 H]C([2 H])([2 H])SCCS(=O)(=O)CCSC([2 H])([2 H])[2 H] |
| IUPAC Name | 1-(trideuteriomethylsulfanyl)-2-[2-(trideuteriomethylsulfanyl)ethylsulfonyl]ethane |
| Molecular Weight (g/mol) | 220.4061 |
| Formula Weight | 220.0533 g/mol |
| Show More |