missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,1-Dimethyl-d6-sulfamide, CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | N,N-Dimethylsulfamide-d6 |
| CAS | 1189689-95-9 |
| Molecular Formula | C2 D6 H2 N2 O2 S |
| Quantity | 0.01 g |
| Synonym | N,N-Di(methyl-d3)sulfamide, N,N-Dimethylsulfamide-d6, N,N-Dimethylsulfamide-D6 (N,N-dimethyl-D6), DMSA-d6, Dimethylsulfamide-d6, 1,1-Dimethyl-d6-sulfamide, Trideuterio-[sulfamoyl(trideuteriomethyl)amino]methane |
| InChI Formula | InChI=1 S/C2H8N2O2S/c1-4(2)7(3,5)6/h1-2H3,(H2,3,5,6)/i1D3,2D3 |
| SMILES | [2 H]C([2 H])([2 H])N(C([2 H])([2 H])[2 H])S(=O)(=O)N |
| IUPAC Name | trideuterio-[sulfamoyl(trideuteriomethyl)amino]methane |
| Molecular Weight (g/mol) | 130.199 |
| Formula Weight | 130.068 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction