A&D Offer
Exclusive to New Lab Start-Up Customers
One-Time Offer
Save 15% on A&D Weighing's analytical balances.
A&D Weighing HR Analytical Balance with Internal Calibration
The HR Analytical Balance with patented C-SHS weighing technology provides performance and a lean compact design. Features include:
- Robust construction, strong protection against dust & water
- Built-in internal calibration at the touch of a button (HR-AZ)
- Shock absorber function
- Auto power ON/OFF function
- B5 size footprint for use in narrow spaces
- Reverse backlit LCD display for easy to read weighing results
- SCF – statistical calculation function
- Standard RS-232C
- Fast compact SHS sensor, response time of about 2 seconds
- USB – quick interface with no driver installation required (optional)
- LAN – ethernet interface with WinCT-Plus software (optional)
- Easily removable large breeze break
- Built-in rechargeable NiMH battery pack (optional)
Fill out the form below to benefit from this offer and to receive the promotion code.
Information About the A&D Weighing HR Analytical Balances with Internal Calibration That You Can Select
Catalog No. 16671182
Capacity: 102 g, Model: HR-100AZ
Catalog No. 16621192
Capacity: 252 g, Model: HR-250AZ