Learn More
A&D Weighing HR Analytical Balance Internal Calibration
HR Analytical Balance with C-SHS patented weighing technology, which provides performance and a lean compact design.
1248.15€ - 1473.14€
Specifications
Display Type | Reverse-backlit LCD |
---|---|
Material | Stainless Steel |
Stabilization Time | 2 sec. |
Electrical Requirements | 11 VA to the AC adaptor (12 V, 1 A) |
Product Code | Brand | Capacity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity | Price | Quantity & Availability | |||||
16621192
|
A&D Weighing
HR-250AZ |
252 g |
1248.15€
Each |
Estimated Shipment: 26-07-2024
Log in to see stock. |
|||||
16641192
|
A&D Weighing
HR-251AZ |
62 g/252 g |
1414.21€
Each |
Estimated Shipment: 26-07-2024
Log in to see stock. |
|||||
16671182
|
A&D Weighing
HR-100AZ |
102 g |
1408.32€
Each |
Estimated Shipment: 26-07-2024
Log in to see stock. |
|||||
16691182
|
A&D Weighing
HR-150AZ |
152 g |
1473.14€
Each |
Estimated Shipment: 26-07-2024
Log in to see stock. |
|||||
Description
- Robust construction, strong protection against dust & water
- Built-in internal calibration at the touch of a button (HR-AZ)
- Shock absorber function
- Auto power ON/OFF function
- B5 size footprint for use in narrow space
- Reverse backlit LCD display for easy to read weighing results
- SCF – Statistical calculation function
- Standard RS-232C
- Fast compact SHS sensor, response time of about 2 seconds
- Quick USB interface – no driver installation required (optional)
- LAN – Ethernet interface with WinCT-Plus software (optional)
- Easily removable large breeze break
- Built-in rechargeable NiMH battery pack (optional)
Specifications
Reverse-backlit LCD | |
Stainless Steel | |
2 sec. | |
11 VA to the AC adaptor (12 V, 1 A) |