accessibility menu, dialog, popup

UserName

CAS RN 97023-48-8

OOH

CAS RN 97023-48-8

IUPAC Name: 2-cyclohexylbenzoic acid
Synonyms: 2-cyclohexylbenzoic acid
Molecular Weight (g/mol): 204.27
Molecular Formula: C13H16O2
InChi Key: ZKTFZNPTAJIXMK-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC=C1C1CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results