CAS RN 96-66-2
CAS RN 96-66-2
IUPAC Name:
2-tert-butyl-4-[(3-tert-butyl-4-hydroxy-5-methylphenyl)sulfanyl]-6-methylphenol
Synonyms:
2-tert-butyl-4-[(3-tert-butyl-4-hydroxy-5-methylphenyl)sulfanyl]-6-methylphenol
Molecular Weight (g/mol):
358.54
Molecular Formula:
C22H30O2S
InChi Key:
YFHKLSPMRRWLKI-UHFFFAOYSA-N
SMILES:
CC1=CC(SC2=CC(C)=C(O)C(=C2)C(C)(C)C)=CC(=C1O)C(C)(C)C
1
–
3
of
3
results
1
4,4'-Thiobis(6-tert-butyl-o-cresol), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
4,4'-Thiobis(6-tert-butyl-o-cresol), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
4,4'-Thiobis(6-tert-butyl-o-cresol), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.