accessibility menu, dialog, popup

UserName

CAS RN 959-36-4

OHNNHO(E)(E)

CAS RN 959-36-4

IUPAC Name: (6E)-6-[(2-{[(1E)-6-oxocyclohexa-2,4-dien-1-ylidene]methyl}hydrazin-1-yl)methylidene]cyclohexa-2,4-dien-1-one
Synonyms: (6E)-6-[(2-{[(1E)-6-oxocyclohexa-2,4-dien-1-ylidene]methyl}hydrazin-1-yl)methylidene]cyclohexa-2,4-dien-1-one
Molecular Weight (g/mol): 240.26
Molecular Formula: C14H12N2O2
InChi Key: SPEXYYIULCBQJR-WGDLNXRISA-N
SMILES: O=C1C=CC=C\C1=C/NN\C=C1/C=CC=CC1=O
Molecular Weight (g/mol) 
  • (8)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (8)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)
  • (3)