CAS RN 948051-93-2
CAS RN 948051-93-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
3-[1-(Methylamino)ethyl]phenyl N-ethyl-N-methylcarbamate, TRC
CAS: 948051-93-2 Molecular Formula: C13 H20 N2 O2 Molecular Weight (g/mol): 236.31 Synonym: Carbamic acid, N-ethyl-N-methyl-, 3-[1-(methylamino)ethyl]phenyl ester,3-[1-(Methylamino)ethyl]phenyl N-ethyl-N-methylcarbamate,Rivastigmine Tartrate Imp E (EP) as Racemate IUPAC Name: [3-[1-(methylamino)ethyl]phenyl] N-ethyl-N-methylcarbamate SMILES: CCN(C)C(=O)Oc1cccc(c1)C(C)NC
2
3-[(1RS)-1-(Methylamino)ethyl]phenyl N-Ethyl-N-methylcarbamate, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.