accessibility menu, dialog, popup

UserName

H3CNOOCH3

CAS RN 947935-14-0

IUPAC Name: N-cyclohexyl-2-methoxy-N-methylbenzamide
Synonyms: N-cyclohexyl-2-methoxy-N-methylbenzamide
Molecular Weight (g/mol): 247.34
Molecular Formula: C15H21NO2
InChi Key: XOFREWHXNLECSD-UHFFFAOYSA-N
SMILES: COC1=CC=CC=C1C(=O)N(C)C1CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results