accessibility menu, dialog, popup

UserName

FNFFF

CAS RN 94239-04-0

IUPAC Name: 2-fluoro-6-(trifluoromethyl)pyridine
Synonyms: 2-fluoro-6-(trifluoromethyl)pyridine
Molecular Weight (g/mol): 165.09
Molecular Formula: C6H3F4N
InChi Key: IZOIOCQPMHHDHN-UHFFFAOYSA-N
SMILES: FC1=CC=CC(=N1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results