CAS RN 94-24-6
CAS RN 94-24-6
IUPAC Name:
2-(dimethylamino)ethyl 4-(butylamino)benzoate
Synonyms:
tetracaine intercain tetracaine preparation medihaler-tetracaine tetracaine (substance) uromucaesthin meethobalm dicaine tetracaina pontocaine
Molecular Weight (g/mol):
264.37
Molecular Formula:
C15H24N2O2
InChi Key:
GKCBAIGFKIBETG-UHFFFAOYSA-N
SMILES:
CCCCNC1=CC=C(C=C1)C(=O)OCCN(C)C
1
–
3
of
3
results
1
Tetracaine, 98%
CAS: 94-24-6 Molecular Formula: C15H24N2O2 Molecular Weight (g/mol): 264.36 InChI Key: GKCBAIGFKIBETG-UHFFFAOYSA-N PubChem CID: 5411 ChEBI: CHEBI:9468
2
Tetracaine, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3
Tetracaine, LoGiCal
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.