accessibility menu, dialog, popup

UserName

CAS RN 94-12-2

H3COONH2

CAS RN 94-12-2

IUPAC Name: propyl 4-aminobenzoate
Synonyms: risocaine risocaina risocainum keloform P raythesin 利索卡因 propyl-P-aminobenzoate propesin (6CI) propesine propylcain
Molecular Weight (g/mol): 179.22
Molecular Formula: C10H13NO2
InChi Key: NBFQYHKHPBMJJV-UHFFFAOYSA-N
SMILES: CCCOC(=O)C1=CC=C(N)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results