CAS RN 93892-40-1
CAS RN 93892-40-1
IUPAC Name:
1,1,3,3-tetramethyl-4,6-bis(propan-2-yl)-2,3-dihydro-1H-inden-5-ol
Synonyms:
4,6-diisopropyl-1,1,3,3-tetramethyl-2H-inden-5-ol
Molecular Weight (g/mol):
274.45
Molecular Formula:
C19H30O
InChi Key:
WGQDDNQFNLMBOS-UHFFFAOYSA-N
SMILES:
CC(C)C1=CC2=C(C(C(C)C)=C1O)C(C)(C)CC2(C)C
1
–
1
of
1
results
1
4,6-Diisopropyl-1,1,3,3-tetramethyl-5-indanol, 97%, Thermo Scientific™
CAS: 93892-40-1 Molecular Formula: C19H30O Molecular Weight (g/mol): 274.448 MDL Number: MFCD00061045 InChI Key: WGQDDNQFNLMBOS-UHFFFAOYSA-N Synonym: 4,6-diisopropyl-1,1,3,3-tetramethyl-5-indanol,4,6-bis isopropyl-1,1,3,3-tetramethylindan-5-ol,4,6-diisopropyl-1,1,3,3-tetramethyl-2h-inden-5-ol,4,6-bis methylethyl-1,1,3,3-tetramethylindan-5-ol,1,1,3,3-tetramethyl-4,6-di propan-2-yl-2h-inden-5-ol,1h-inden-5-ol, 2,3-dihydro-1,1,3,3-tetramethyl-4,6-bis 1-methylethyl,2,3-dihydro-1,1,3,3-tetramethyl-4,6-bis 1-methylethyl-1h-inden-5-ol PubChem CID: 621162 IUPAC Name: 1,1,3,3-tetramethyl-4,6-di(propan-2-yl)-2H-inden-5-ol SMILES: CC(C)C1=C(C(=C2C(=C1)C(CC2(C)C)(C)C)C(C)C)O