accessibility menu, dialog, popup

UserName

CAS RN 92973-26-7

ClOOCl

CAS RN 92973-26-7

IUPAC Name: 5-(3-chlorophenyl)furan-2-carbonyl chloride
Synonyms: 5-(3-chlorophenyl)furan-2-carbonyl chloride
Molecular Weight (g/mol): 241.07
Molecular Formula: C11H6Cl2O2
InChi Key: DBONYOXUBWINEQ-UHFFFAOYSA-N
SMILES: ClC(=O)C1=CC=C(O1)C1=CC(Cl)=CC=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results