accessibility menu, dialog, popup

UserName

CAS RN 926247-77-0

SH2NOFFF

CAS RN 926247-77-0

IUPAC Name: 2-(trifluoromethoxy)benzene-1-carbothioamide
Synonyms: 2-(trifluoromethoxy)benzenecarbothioamide
Molecular Weight (g/mol): 221.2
Molecular Formula: C8H6F3NOS
InChi Key: WVRLRVCJDVSKRC-UHFFFAOYSA-N
SMILES: NC(=S)C1=CC=CC=C1OC(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results