accessibility menu, dialog, popup

UserName

CAS RN 916420-66-1

ClH2NFCl

CAS RN 916420-66-1

IUPAC Name: 1-(3,6-dichloro-2-fluorophenyl)methanamine
Synonyms: 1-(3,6-dichloro-2-fluorophenyl)methanamine
Molecular Weight (g/mol): 194.03
Molecular Formula: C7H6Cl2FN
InChi Key: CSAKSJHNYMDUQJ-UHFFFAOYSA-N
SMILES: NCC1=C(Cl)C=CC(Cl)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results