CAS RN 914981-47-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Fmoc-aminopropyloxy-3-DMT-glycerol, TRC
CAS: 914981-47-8 Molecular Formula: C42 H43 N O7 Molecular Weight (g/mol): 673.79 Synonym: (9H-Fluoren-9-yl)methyl (3-(3-(Bis(4-methoxyphenyl)(phenyl)methoxy)-2-hydroxypropoxy)propyl)carbamate IUPAC Name: 9H-fluoren-9-ylmethyl N-[3-[3-[bis(4-methoxyphenyl)-phenyl-methoxy]-2-hydroxy-propoxy]propyl]carbamate SMILES: O=C(NCCCOCC(O)COC(C1=CC=C(OC)C=C1)(C2=CC=CC=C2)C3=CC=C(OC)C=C3)OCC4C5=C(C=CC=C5)C6=C4C=CC=C6