accessibility menu, dialog, popup

UserName

CH3OBrFFF

CAS RN 914635-58-8

IUPAC Name: 4-bromo-1-ethoxy-2-(trifluoromethyl)benzene
Synonyms: 4-bromo-1-ethoxy-2-(trifluoromethyl)benzene
Molecular Weight (g/mol): 269.06
Molecular Formula: C9H8BrF3O
InChi Key: BGDWEDIMCBCUDB-UHFFFAOYSA-N
SMILES: CCOC1=C(C=C(Br)C=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results