accessibility menu, dialog, popup

UserName

CAS RN 91374-20-8

HClCH3NH3CNHO

CAS RN 91374-20-8

IUPAC Name: hydrogen 4-[2-(dipropylamino)ethyl]-2,3-dihydro-1H-indol-2-one chloride
Synonyms: ropinirole
Molecular Weight (g/mol): 296.84
Molecular Formula: C16H25ClN2O
InChi Key: XDXHAEQXIBQUEZ-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CCCN(CCC)CCC1=CC=CC2=C1CC(=O)N2
Certifications/Compliance 
  • (1)
  • (1)
Industry Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results