accessibility menu, dialog, popup

UserName

CAS RN 9003-20-7

OOMen

CAS RN 9003-20-7

IUPAC Name: ethenyl acetate
Synonyms: vinyl acetate acetate de vinyle zeset T vinylacetat vinyl A monomer vinylazetat vinile (acetato di) vinyl acetate monomer vinyl acetate H.Q. essigsaeurevinylester
Molecular Weight (g/mol): 86.09
Molecular Formula: C4H6O2
InChi Key: XTXRWKRVRITETP-UHFFFAOYSA-N
SMILES: CC(=O)OC=C
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)