accessibility menu, dialog, popup

UserName

CAS RN 89-05-4

HOOOHOOHOOHO

CAS RN 89-05-4

IUPAC Name: benzene-1,2,4,5-tetracarboxylic acid
Synonyms: pyromellitic acid monosodium salt pyromellitic acid pyromellitic acid, thorium (4+) salt pyromellitic acid, maganese salt tetrasilver (1+) salt pyromellitic acid cesium salt pyromellitic acid pyromellitic acid, tetrasilver (1+) salt copper (2+) salt pyromellitic acid usaf XR-20 pyromellitic acid, cobalt salt
Molecular Weight (g/mol): 254.15
Molecular Formula: C10H6O8
InChi Key: CYIDZMCFTVVTJO-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(C(O)=O)=C(C=C1C(O)=O)C(O)=O
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)