accessibility menu, dialog, popup

UserName

FFFHOF

CAS RN 886761-79-1

IUPAC Name: 2-[2-fluoro-5-(trifluoromethyl)phenyl]ethan-1-ol
Synonyms: 2-[2-fluoro-5-(trifluoromethyl)phenyl]ethanol
Molecular Weight (g/mol): 208.16
Molecular Formula: C9H8F4O
InChi Key: YYIXBCMBMSZIOV-UHFFFAOYSA-N
SMILES: OCCC1=C(F)C=CC(=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results