accessibility menu, dialog, popup

UserName

CAS RN 886499-88-3

BrHOFCl

CAS RN 886499-88-3

IUPAC Name: 2-bromo-4-chloro-6-fluorophenol
Synonyms: 2-bromo-4-chloro-6-fluorophenol
Molecular Weight (g/mol): 225.44
Molecular Formula: C6H3BrClFO
InChi Key: VGDYUSDMWUDGAB-UHFFFAOYSA-N
SMILES: OC1=C(Br)C=C(Cl)C=C1F
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results