CAS RN 88462-65-1
IUPAC Name:
1-[3-(trifluoromethyl)phenoxy]but-3-yn-2-ol
Synonyms:
1-[3-(trifluoromethyl)phenoxy]but-3-yn-2-ol
Molecular Weight (g/mol):
230.19
Molecular Formula:
C11H9F3O2
InChi Key:
OPWHACCQYXGLRX-UHFFFAOYNA-N
SMILES:
OC(COC1=CC=CC(=C1)C(F)(F)F)C#C
1
–
1
of
1
results
1
1-(3-Trifluoromethylphenoxy)-3-butyn-2-ol, 98%
CAS: 88462-65-1 Molecular Formula: C11H9F3O2 Molecular Weight (g/mol): 230.19 MDL Number: MFCD02683085 InChI Key: OPWHACCQYXGLRX-UHFFFAOYNA-N Synonym: 1-3-trifluoromethylphenoxy-3-butyn-2-ol,1-3-trifluoromethyl phenoxy but-3-yn-2-ol,3-butyn-2-ol,1-3-trifluoromethyl phenoxy,acmc-20drht,1-3-trifluoromethylphenoxy-3-butyn-2-o PubChem CID: 11172256 IUPAC Name: 1-[3-(trifluoromethyl)phenoxy]but-3-yn-2-ol SMILES: OC(COC1=CC=CC(=C1)C(F)(F)F)C#C