accessibility menu, dialog, popup

UserName

FFFFFFFFFFFFOH

CAS RN 87901-76-6

IUPAC Name: bis[3,5-bis(trifluoromethyl)phenyl]methanol
Synonyms: bis[3,5-bis(trifluoromethyl)phenyl]methanol
Molecular Weight (g/mol): 456.23
Molecular Formula: C17H8F12O
InChi Key: DUCQGUCBWHWEBT-UHFFFAOYSA-N
SMILES: OC(C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results